Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018448.4 | NP_060918.2 | 2 | 634 | ||
GenBank mRNA | AB020636.2 | - | 2 | 536 | ||
AK096010.1 | - | 2 | 540 | |||
AK292456.1 | - | 4 | 473 | |||
AK314358.1 | - | 2 | 522 | |||
AL136810.1 | - | 2 | 546 | |||
BC050341.1 | - | 2 | 330 | 5265409 |