Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018449.3 | NP_060919.3 | 2, 3 | 220 | ||
GenBank mRNA | AB209911.1 | - | 2, 3 | 196 | ||
AF157327.1 | - | 1, 2 | 133 | |||
AK302335.1 | - | 2, 3 | 200 | |||
BG723831.1 | - | 2, 3 | 226 |