Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320041.1 | NP_001306970.1 | 2 | 259 | ||
NM_018456.5 | NP_060926.2 | 2 | 259 | |||
XM_005247618.3 | XP_005247675.1 | 3 | 409 | |||
XM_017006861.1 | XP_016862350.1 | 3 | 131 | |||
XM_017006862.1 | XP_016862351.1 | 2 | 168 | |||
XM_017006863.1 | XP_016862352.1 | 2 | 202 | |||
GenBank mRNA | AF517829.1 | - | 2 | 204 | ||
AK301560.1 | - | 2 | 202 | |||
AY049020.1 | - | 2 | 224 | |||
BC014209.2 | - | 2 | 219 | 4765823 | ||
BU941228.1 | - | 255 | ||||
HY005912.1 | - | 2 | 260 |