Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CISD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018464.4 | NP_060934.1 | 2 | 369 | ||
XM_017016409.1 | XP_016871898.1 | 3 | 610 | |||
GenBank mRNA | AF220049.1 | - | 2 | 308 | ||
AK312017.1 | - | 2 | 244 | |||
AY960578.1 | - | 2 | 252 | |||
BC005962.1 | - | 2 | 345 | 4051044 | ||
BC007043.1 | - | 2 | 301 | 3930701 | ||
BC008474.1 | - | 2 | 306 | 4283253 | ||
BC059168.1 | - | 2 | 316 | 4132410 | ||
DA417986.1 | - | 2 | 369 |