Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLGRKT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018465.3 | NP_060935.2 | 4 | 378 | ||
XM_005251510.4 | XP_005251567.1 | 4 | 493 | |||
XM_005251512.4 | XP_005251569.1 | 2 | 166 | |||
XM_011517960.2 | XP_011516262.1 | 4 | 526 | |||
GenBank mRNA | AF220050.1 | - | 4 | 302 | ||
AF225420.1 | - | 4 | 319 | |||
AK027594.1 | - | 4 | 317 | |||
AK312736.1 | - | 4 | 307 | |||
BC008212.1 | - | 4 | 288 | 3871917 | ||
BM693747.1 | - | 4 | 378 |