Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080500.2 | NP_001073969.1 | 2 | 495 | ||
GenBank mRNA | AB374231.1 | - | 1 | 365 | ||
BC146903.1 | - | 2 | 495 | 9056831 | ||
BC146911.1 | - | 2 | 495 | 9056839 | ||
BC146931.1 | - | 2 | 495 | 9056859 | ||
BC146935.1 | - | 2 | 495 | 9056863 | ||
BC171841.1 | - | 2 | 495 | 9054495 | ||
EU541473.1 | - | 1 | 368 |