Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200049.2 | NP_001186978.2 | 38 | 5455 | ||
GenBank mRNA | DV080461.1 | - | 3 | 340 |