Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267571.1 | NP_001254500.1 | 8 | 1703 | ||
NM_001267572.1 | NP_001254501.1 | 2 | 482 | |||
NM_018421.3 | NP_060891.3 | 8 | 1703 | |||
NR_051978.1 | 7 | 1561 | ||||
XM_011518843.1 | XP_011517145.1 | 3 | 1585 | |||
XM_017014899.1 | XP_016870388.1 | 8 | 1133 | |||
GenBank mRNA | AB449882.1 | - | 8 | 1523 | ||
AF318370.1 | - | 8 | 1562 | |||
AK026105.1 | - | 7 | 1051 | |||
AK124772.1 | - | 7 | 2252 | |||
AK131290.1 | - | 8 | 1581 | |||
AK298575.1 | - | 9 | 1353 | |||
AL353935.1 | - | 3 | 1764 | |||
AY026527.1 | - | 8 | 1615 | |||
BC028918.1 | - | 2 | 482 | 4931447 | ||
BC071978.1 | - | 4 | 590 | 5443205 |