Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282529.1 | NP_001269458.1 | 3 | 227 | ||
NM_018449.3 | NP_060919.3 | 4 | 314 | |||
GenBank mRNA | AB209911.1 | - | 3 | 2743 | ||
AF157327.1 | - | 3 | 227 | |||
AK302335.1 | - | 4 | 294 | |||
AK302754.1 | - | 5 | 341 | |||
AK303493.1 | - | 3 | 165 | |||
BG723831.1 | - | 4 | 320 |