Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018452.5 | NP_060922.2 | 4 | 1173 | ||
GenBank mRNA | AF161405.1 | - | 4 | 510 | ||
AF217510.1 | - | 4 | 516 | |||
AK000891.1 | - | 4 | 421 | |||
BC029130.1 | - | 4 | 441 | 5182086 | ||
BC146893.1 | - | 4 | 483 | 9056821 | ||
BC146895.1 | - | 4 | 483 | 9056823 |