Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EAPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318916.1 | NP_001305845.1 | 2 | 348 | ||
NM_018453.3 | NP_060923.2 | 2 | 348 | |||
XM_011536958.1 | XP_011535260.1 | 2 | 626 | |||
XM_017021460.1 | XP_016876949.1 | 2 | 348 | |||
GenBank mRNA | AF217512.1 | - | 2 | 334 | ||
AK000585.1 | - | 2 | 312 | |||
AK225467.1 | - | 2 | 303 | |||
AK225477.1 | - | 2 | 312 | |||
AK225991.1 | - | 2 | 312 | |||
AV757492.1 | - | 2 | 433 | |||
AY157300.1 | - | 2 | 330 | |||
AY157301.1 | - | 2 | 330 | |||
AY869694.1 | - | 2 | 271 | |||
BC001245.1 | - | 2 | 321 | 3460193 | ||
BU662131.1 | - | 2 | 296 | |||
CD702290.1 | - | 2 | 377 |