Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005272464.4 | XP_005272521.1 | 7 | 939 | ||
XM_006722705.3 | XP_006722768.1 | 7 | 1071 | |||
XM_006722706.3 | XP_006722769.1 | 8 | 1234 | |||
XM_011527835.2 | XP_011526137.1 | 7 | 939 | |||
XM_011527836.2 | XP_011526138.1 | 7 | 939 | |||
XM_011527837.2 | XP_011526139.1 | 7 | 939 | |||
XM_011527838.2 | XP_011526140.1 | 6 | 627 | |||
XM_011527839.2 | XP_011526141.1 | 6 | 693 | |||
XM_011527841.2 | XP_011526143.1 | 7 | 932 |