Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103091866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111954.1 | 4 | 949 | |||
NR_111955.1 | 2 | 616 | ||||
NR_111956.1 | 3 | 705 | ||||
NR_111957.1 | 4 | 915 | ||||
NR_111958.1 | 3 | 860 | ||||
GenBank mRNA | AK307112.1 | - | 3 | 639 | ||
AK309334.1 | - | 4 | 658 | |||
CA777704.1 | - | 4 | 664 |
Gene Symbol: LOC388692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124550.1 | - | 1 | 1192 |