Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110630.1 | 3 | 385 | |||
GenBank mRNA | DB443056.1 | - | 3 | 385 |