Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGPAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018361.3 | NP_060831.2 | 8 | 2965 | ||
XM_011534751.2 | XP_011533053.1 | 6 | 2485 | |||
GenBank mRNA | AK002072.1 | - | 8 | 2629 | ||
AL136587.1 | - | 8 | 2655 | |||
AL514578.3 | - | 1 | 278 | |||
BC068519.1 | - | 8 | 2628 |