Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145122.1 | NP_001138594.1 | 8 | 958 | ||
NM_001321270.1 | NP_001308199.1 | 8 | 1022 | |||
XM_011532863.2 | XP_011531165.1 | 9 | 1119 | |||
XM_011532864.2 | XP_011531166.1 | 9 | 1156 | |||
XM_011532865.1 | XP_011531167.1 | 9 | 1060 | |||
XM_011532866.2 | XP_011531168.1 | 7 | 827 | |||
GenBank mRNA | AK092257.1 | - | 8 | 923 | ||
AK296910.1 | - | 10 | 926 |