Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001370.1 | NP_001361.1 | 25 | 3979 | ||
XM_006711956.2 | XP_006712019.1 | 25 | 4084 | |||
XM_011532649.2 | XP_011530951.1 | 25 | 3933 | |||
XM_011532650.2 | XP_011530952.1 | 25 | 3934 | |||
XM_011532652.1 | XP_011530954.1 | 25 | 3979 | |||
XM_011532653.1 | XP_011530955.1 | 25 | 3979 | |||
XM_011532654.1 | XP_011530956.1 | 24 | 3712 | |||
XM_011532655.2 | XP_011530957.1 | 21 | 3001 | |||
XM_011532657.1 | XP_011530959.1 | 19 | 2740 | |||
XM_011532660.1 | XP_011530962.1 | 25 | 3979 | |||
XM_017003521.1 | XP_016859010.1 | 25 | 3979 | |||
XM_017003522.1 | XP_016859011.1 | 20 | 3805 | |||
XM_017003523.1 | XP_016859012.1 | 20 | 2972 | |||
XM_017003524.1 | XP_016859013.1 | 25 | 3979 |