Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HJURP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282962.1 | NP_001269891.1 | 6 | 823 | ||
NM_001282963.1 | NP_001269892.1 | 5 | 730 | |||
NM_018410.4 | NP_060880.3 | 8 | 985 | |||
XM_011511437.1 | XP_011509739.1 | 7 | 889 | |||
GenBank mRNA | AB101211.1 | - | 8 | 986 | ||
AB162218.1 | - | 8 | 919 | |||
AK074809.1 | - | 8 | 951 | |||
AK301643.1 | - | 5 | 699 | |||
AK303109.1 | - | 6 | 764 | |||
AL162048.1 | - | 1 | 162 | |||
BC001940.1 | - | 8 | 919 | 2820741 | ||
BM475244.1 | - | 1 | 243 | |||
DN998761.1 | - | 3 | 537 |