Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018420.2 | NP_060890.2 | 12 | 2741 | ||
GenBank mRNA | AK055633.1 | - | 1 | 785 | ||
AK125072.1 | - | 10 | 2310 | |||
AL353933.1 | - | 4 | 1442 | |||
BC026358.1 | - | 12 | 2740 | 4814675 |