Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138572.2 | NP_612639.2 | 9 | 2201 | ||
XM_011514296.1 | XP_011512598.1 | 10 | 2314 | |||
XM_017010243.1 | XP_016865732.1 | 9 | 2355 | |||
XM_017010245.1 | XP_016865734.1 | 7 | 1899 | |||
GenBank mRNA | AF086309.1 | - | 1 | 444 | ||
AL833401.1 | - | 10 | 2313 |