Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282529.1 | NP_001269458.1 | 11 | 1398 | ||
NM_018449.3 | NP_060919.3 | 15 | 1712 | |||
GenBank mRNA | AB040924.1 | - | 3 | 514 | ||
AK025801.1 | - | 4 | 602 | |||
AK026088.1 | - | 7 | 880 | |||
AK074884.1 | - | 2 | 71 | |||
AK294952.1 | - | 15 | 1630 | |||
AK299183.1 | - | 9 | 1120 | |||
AK302335.1 | - | 13 | 1533 | |||
AK302754.1 | - | 14 | 1580 | |||
AK303493.1 | - | 11 | 1336 | |||
AY358682.1 | - | 3 | 922 | |||
BC111762.1 | - | 9 | 1082 |