Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114118.2 | NP_001107590.1 | 10 | 1056 | ||
XM_005256717.2 | XP_005256774.1 | 10 | 1056 | |||
XM_006721556.2 | XP_006721619.1 | 9 | 945 | |||
XM_011523954.2 | XP_011522256.1 | 10 | 973 | |||
XM_011523955.2 | XP_011522257.1 | 9 | 827 | |||
XM_017024826.1 | XP_016880315.1 | 9 | 943 | |||
XM_017024827.1 | XP_016880316.1 | 4 | 530 | |||
XM_017024828.1 | XP_016880317.1 | 5 | 348 | |||
XM_017024829.1 | XP_016880318.1 | 5 | 348 | |||
GenBank mRNA | AJ277841.1 | - | 5 | 353 | ||
AK125048.1 | - | 7 | 1745 | |||
AK223470.1 | - | 3 | 232 | |||
BC010707.1 | - | 3 | 171 | |||
BP344033.1 | - | 4 | 305 |