Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018646.5 | NP_061116.5 | 9, 10 | 1446 | ||
GenBank mRNA | AF304463.1 | - | 9, 10 | 1217 | ||
AF365927.1 | - | 9, 10 | 1433 | |||
AF365928.1 | - | 5, 6 | 860 | |||
AJ243500.2 | - | 9, 10 | 1417 | |||
AJ243501.2 | - | 9, 10 | 1417 | |||
AJ487964.2 | - | 10, 11 | 1386 | |||
AK075103.1 | - | 4, 5 | 588 | |||
AK291707.1 | - | 9, 10 | 1447 | |||
AY203947.1 | - | 6, 7 | 1627 | |||
KF534785.1 | - | 9, 10 | 1446 |