Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015203.1 | XP_016870692.1 | 1 | 215 | ||
XM_017015204.1 | XP_016870693.1 | 1 | 215 | |||
XM_017015205.1 | XP_016870694.1 | 1 | 215 |