Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGAPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020343.3 | NP_065076.2 | 20 | 2838 | ||
XM_005260768.2 | XP_005260825.1 | 20 | 2853 | |||
XM_006723598.1 | XP_006723661.1 | 21 | 2994 | |||
XM_006723599.1 | XP_006723662.1 | 21 | 2994 | |||
XM_011529309.1 | XP_011527611.1 | 21 | 2994 | |||
XM_011529310.2 | XP_011527612.1 | 19 | 2615 | |||
XM_011529311.2 | XP_011527613.2 | 7 | 2954 | |||
XM_017027972.1 | XP_016883461.1 | 21 | 2994 | |||
XM_017027973.1 | XP_016883462.1 | 18 | 2457 | |||
XM_017027974.1 | XP_016883463.1 | 18 | 2452 | |||
XM_017027975.1 | XP_016883464.1 | 20 | 2786 | |||
XM_017027976.1 | XP_016883465.1 | 18 | 2639 | |||
XM_017027977.1 | XP_016883466.1 | 21 | 2994 | |||
GenBank mRNA | AK002211.1 | - | 12 | 1708 | ||
BC171842.1 | - | 18 | 2639 | |||
DQ310704.1 | - | 20 | 2695 |