Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080428.2 | NP_001073897.2 | 24 | 6876 | ||
NM_001122679.1 | NP_001116151.1 | 28 | 7448 | |||
XM_005265950.2 | XP_005266007.1 | 30 | 8095 | |||
XM_005265952.2 | XP_005266009.1 | 26 | 6822 | |||
XM_006714897.2 | XP_006714960.1 | 29 | 8074 | |||
XM_011534604.2 | XP_011532906.1 | 29 | 7909 | |||
XM_017009660.1 | XP_016865149.1 | 30 | 8122 | |||
XM_017009661.1 | XP_016865150.1 | 29 | 8101 | |||
XM_017009662.1 | XP_016865151.1 | 29 | 7912 | |||
XM_017009663.1 | XP_016865152.1 | 27 | 7317 | |||
XM_017009664.1 | XP_016865153.1 | 27 | 7230 | |||
XM_017009665.1 | XP_016865154.1 | 27 | 7026 | |||
XM_017009666.1 | XP_016865155.1 | 26 | 7005 | |||
XM_017009667.1 | XP_016865156.1 | 27 | 6999 | |||
XM_017009668.1 | XP_016865157.1 | 27 | 8536 | |||
XM_017009669.1 | XP_016865158.1 | 27 | 8368 | |||
GenBank mRNA | AB032953.2 | - | 19 | 5585 | ||
AK293262.1 | - | 2 | 307 | |||
AK302302.1 | - | 2 | 1013 | |||
AL137500.1 | - | 3 | 1701 | |||
BX648178.1 | - | 23 | 6470 | |||
CR936703.1 | - | 24 | 6809 |