Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRAMD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286563.1 | NP_001273492.1 | 1, 2 | 528 | ||
NM_020716.2 | NP_065767.1 | 1, 2 | 528 | |||
XM_005271619.4 | XP_005271676.1 | 1, 2 | 528 | |||
XM_017018044.1 | XP_016873533.1 | 1, 2 | 528 | |||
GenBank mRNA | AK127457.1 | - | 1, 2 | 528 | ||
BC105741.1 | - | 1, 2 | 342 | 40026783 | ||
BC105931.1 | - | 1, 2 | 342 | 40026781 | ||
BC105932.1 | - | 1, 2 | 342 | 40026782 | ||
BC107480.1 | - | 1, 2 | 342 | 40026784 |