Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020780.1 | NP_065831.1 | 2 | 464 | ||
XM_011541828.2 | XP_011540130.1 | 1 | 2566 | |||
XM_011541830.2 | XP_011540132.1 | 2 | 547 | |||
XM_011541831.2 | XP_011540133.1 | 2 | 547 | |||
XM_011541832.2 | XP_011540134.1 | 2 | 547 | |||
XM_011541833.2 | XP_011540135.1 | 2 | 547 | |||
XM_011541835.2 | XP_011540137.1 | 2 | 547 | |||
GenBank mRNA | AB037758.1 | - | 2 | 464 | ||
AB593117.1 | - | 2 | 547 |