Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285549.1 | NP_001272478.1 | 7 | 5494 | ||
NM_020923.2 | NP_065974.1 | 5 | 5601 | |||
XM_005246711.3 | XP_005246768.1 | 6 | 5601 | |||
XM_005246712.4 | XP_005246769.1 | 6 | 5424 | |||
XM_005246713.3 | XP_005246770.1 | 6 | 5418 | |||
XM_006712656.3 | XP_006712719.1 | 6 | 5457 | |||
XM_011511532.2 | XP_011509834.1 | 6 | 5634 | |||
XM_011511533.2 | XP_011509835.1 | 6 | 5451 | |||
XM_011511534.2 | XP_011509836.1 | 5 | 5374 | |||
XM_011511535.2 | XP_011509837.1 | 5 | 5380 | |||
XM_017004573.1 | XP_016860062.1 | 7 | 5707 | |||
XM_017004574.1 | XP_016860063.1 | 7 | 5633 | |||
XM_017004575.1 | XP_016860064.1 | 6 | 5602 | |||
XM_017004576.1 | XP_016860065.1 | 2 | 3603 | |||
GenBank mRNA | AB046791.2 | - | 1 | 3469 | ||
AB775780.1 | - | 7 | 5495 | |||
AB775781.1 | - | 5 | 5601 | |||
AK130893.1 | - | 1 | 1844 |