Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021623.1 | NP_067636.1 | 11 | 1105 | ||
XM_011544605.2 | XP_011542907.1 | 11 | 990 | |||
XM_011544606.2 | XP_011542908.1 | 11 | 1041 | |||
XM_011544607.2 | XP_011542909.1 | 11 | 1003 | |||
XM_017013718.1 | XP_016869207.1 | 10 | 856 | |||
XM_017013719.1 | XP_016869208.1 | 10 | 826 | |||
XM_017013720.1 | XP_016869209.1 | 10 | 941 | |||
GenBank mRNA | AF286164.1 | - | 11 | 1030 | ||
AK291842.1 | - | 11 | 1049 | |||
AK296068.1 | - | 10 | 898 | |||
AK309020.1 | - | 4 | 222 | |||
DC332022.1 | - | 4 | 222 |