Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP7D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173517.1 | NP_001166988.1 | 18 | 3086 | ||
GenBank mRNA | AL832120.1 | - | 18 | 2878 |