Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080457.1 | NP_001073926.1 | 3 | 2230 | ||
XM_005259429.4 | XP_005259486.1 | 3 | 2409 | |||
XM_006723506.3 | XP_006723569.1 | 4 | 2511 | |||
XM_011527520.2 | XP_011525822.1 | 3 | 3657 | |||
GenBank mRNA | BC056207.1 | - | 1 | 437 | ||
BC131897.1 | - | 4 | 1327 |