Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SETD5 SIRNA Assays ›
Gene Symbol:
SETD5
Gene Name:
SET domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.3: 9397700 - 9478154 on Build GRCh38
UniGene ID:
Hs.288164
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
272464
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SETD5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080517.2
NP_001073986.1
12
1851
NM_001292043.1
NP_001278972.1
14
2115
XM_005265301.1
XP_005265358.1
12
1889
XM_005265303.1
XP_005265360.1
14
2270
XM_011533920.1
XP_011532222.1
15
2365
XM_011533921.1
XP_011532223.1
14
1967
XM_011533922.1
XP_011532224.1
13
1928
XM_011533927.1
XP_011532229.1
15
2308
XM_011533928.1
XP_011532230.1
13
1871
XM_011533930.1
XP_011532232.1
15
2309
XM_011533932.1
XP_011532234.1
13
2076
XM_011533933.1
XP_011532235.1
11
1445
XM_017006767.1
XP_016862256.1
15
4027
XM_017006768.1
XP_016862257.1
13
3541
XM_017006769.1
XP_016862258.1
13
5644
XM_017006770.1
XP_016862259.1
14
5683
XM_017006771.1
XP_016862260.1
12
1889
XM_017006772.1
XP_016862261.1
13
5644
XM_017006773.1
XP_016862262.1
13
5587
XM_017006774.1
XP_016862263.1
12
1832
XM_017006775.1
XP_016862264.1
13
5587
XM_017006776.1
XP_016862265.1
14
2172
XM_017006777.1
XP_016862266.1
14
2115
XM_017006778.1
XP_016862267.1
15
2156
XM_017006779.1
XP_016862268.1
13
2133
XM_017006780.1
XP_016862269.1
14
2117
XM_017006781.1
XP_016862270.1
15
2156
XM_017006782.1
XP_016862271.1
14
5683
XM_017006783.1
XP_016862272.1
16
2381
XM_017006784.1
XP_016862273.1
14
5683
XM_017006785.1
XP_016862274.1
14
5683
XM_017006786.1
XP_016862275.1
14
5683
GenBank mRNA
AB051544.1
-
8
1645
AK001569.1
-
1
34
AK091393.1
-
9
1154
AK092046.1
-
14
2070
AK127298.1
-
2
738
AK289529.1
-
3
396
BC020956.1
-
1
65
3851178
BX648380.1
-
4
453
CR627408.1
-
11
1467
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0