Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014247.2 | NP_055062.1 | 10 | 1653 | ||
XM_005263358.3 | XP_005263415.1 | 16 | 1920 | |||
XM_005263359.3 | XP_005263416.1 | 16 | 1922 | |||
XM_005263360.3 | XP_005263417.1 | 16 | 1914 | |||
XM_005263361.3 | XP_005263418.1 | 15 | 1902 | |||
XM_006714420.3 | XP_006714483.1 | 15 | 1902 | |||
XM_006714421.3 | XP_006714484.1 | 16 | 1887 | |||
XM_006714422.3 | XP_006714485.1 | 13 | 2135 | |||
XM_011532425.2 | XP_011530727.1 | 12 | 1750 | |||
XM_011532426.2 | XP_011530728.1 | 13 | 2687 | |||
XM_011532427.2 | XP_011530729.1 | 10 | 2337 | |||
XM_017008856.1 | XP_016864345.1 | 16 | 1921 | |||
XM_017008857.1 | XP_016864346.1 | 13 | 2686 | |||
XM_017008858.1 | XP_016864347.1 | 13 | 1654 | |||
XM_017008859.1 | XP_016864348.1 | 10 | 2337 | |||
GenBank mRNA | AB002311.1 | - | 10 | 1405 | ||
AK294459.1 | - | 10 | 1654 | |||
BC117321.1 | - | 10 | 1331 | 40125872 |