Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015016.1 | NP_055831.1 | 24 | 3011 | ||
XM_005259823.3 | XP_005259880.1 | 24 | 3375 | |||
XM_005259824.2 | XP_005259881.1 | 25 | 3061 | |||
XM_005259825.2 | XP_005259882.1 | 25 | 3058 | |||
XM_005259826.3 | XP_005259883.1 | 23 | 3171 | |||
XM_005259827.2 | XP_005259884.1 | 23 | 3029 | |||
XM_006722698.2 | XP_006722761.1 | 24 | 3053 | |||
XM_006722699.2 | XP_006722762.2 | 24 | 3362 | |||
XM_006722700.3 | XP_006722763.1 | 23 | 3171 | |||
XM_011527823.1 | XP_011526125.1 | 26 | 3151 | |||
XM_017026506.1 | XP_016881995.1 | 24 | 3359 | |||
XM_017026507.1 | XP_016881996.1 | 23 | 3341 | |||
XM_017026508.1 | XP_016881997.1 | 23 | 3339 | |||
XM_017026509.1 | XP_016881998.1 | 24 | 3362 | |||
XM_017026510.1 | XP_016881999.1 | 23 | 3342 | |||
XM_017026511.1 | XP_016882000.1 | 25 | 3449 | |||
XM_017026512.1 | XP_016882001.1 | 24 | 3040 | |||
XM_017026513.1 | XP_016882002.1 | 24 | 3037 | |||
XM_017026514.1 | XP_016882003.1 | 25 | 3044 | |||
XM_017026515.1 | XP_016882004.1 | 24 | 3050 | |||
XM_017026516.1 | XP_016882005.1 | 24 | 3359 | |||
GenBank mRNA | AB011133.1 | - | 24 | 3009 |