Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAGE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018666.2 | NP_061136.2 | 3 | 299 | ||
XM_017029621.1 | XP_016885110.1 | 3 | 307 | |||
XM_017029622.1 | XP_016885111.1 | 3 | 218 | |||
XM_017029623.1 | XP_016885112.1 | 3 | 214 | |||
XM_017029624.1 | XP_016885113.1 | 3 | 214 | |||
XM_017029625.1 | XP_016885114.1 | 3 | 214 | |||
GenBank mRNA | AJ278111.1 | - | 3 | 299 | ||
AK310412.1 | - | 3 | 348 | |||
BC144260.1 | - | 3 | 180 | 9052780 |