Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080439.2 | NP_001073908.2 | 6 | 3176 | ||
XM_011524283.1 | XP_011522585.1 | 6 | 3189 | |||
GenBank mRNA | AL137385.1 | - | 1 | 1203 | ||
BC033017.1 | - | 6 | 3140 | 4825807 | ||
BC033020.1 | - | 6 | 3163 | 4823233 | ||
BC115385.1 | - | 7 | 2752 | 40038134 | ||
BC115386.1 | - | 7 | 2748 |