Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242521.1 | NP_001229450.1 | 3 | 295 | ||
GenBank mRNA | BX377446.2 | - | 3 | 294 |