Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085447.1 | NP_001078916.1 | 3 | 496 | ||
XM_011510589.1 | XP_011508891.1 | 3 | 384 | |||
XM_011510590.1 | XP_011508892.1 | 3 | 417 | |||
GenBank mRNA | AK301851.1 | - | 3 | 464 | ||
BC117445.1 | - | 3 | 439 | 40125996 | ||
BQ432665.1 | - | 3 | 498 |