Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018696.2 | NP_061166.1 | 2, 3 | 271 | ||
GenBank mRNA | AB029151.1 | - | 2, 3 | 211 | ||
AF308695.1 | - | 2, 3 | 255 | |||
AI870985.1 | - | 2, 3 | 272 | |||
AK024822.1 | - | 2, 3 | 251 | |||
AK223507.1 | - | 2, 3 | 215 | |||
AK310911.1 | - | 2, 3 | 240 | |||
BC014624.2 | - | 2, 3 | 232 | 2960155 |