Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100915.1 | NP_001094385.1 | 2 | 90 | ||
GenBank mRNA | AK097481.1 | - | 2 | 93 | ||
AK302567.1 | - | 2 | 85 | |||
AK302748.1 | - | 2 | 74 | |||
BC070103.1 | - | 2 | 58 | |||
BC094790.1 | - | 2 | 61 | |||
BC117335.1 | - | 2 | 90 | 40125886 |