Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100915.1 | NP_001094385.1 | 5 | 768 | ||
GenBank mRNA | AK097481.1 | - | 5 | 771 | ||
AK302567.1 | - | 4 | 1986 | |||
AK302748.1 | - | 5 | 752 | |||
BC070103.1 | - | 5 | 736 | |||
BC117335.1 | - | 5 | 768 | 40125886 |