Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCDC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099434.1 | NP_001092904.1 | 1 | 209 | ||
XM_017000413.1 | XP_016855902.1 | 1 | 490 | |||
GenBank mRNA | BC041458.1 | - | 1 | 207 | ||
BC128073.1 | - | 1 | 209 | 40109068 |