Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCWPW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040432.3 | NP_001035522.1 | 7 | 889 | ||
NM_001324169.1 | NP_001311098.1 | 6 | 769 | |||
NM_001324170.1 | NP_001311099.1 | 5 | 651 | |||
NR_136708.1 | 5 | 651 | ||||
XM_017005755.1 | XP_016861244.1 | 6 | 4564 | |||
XM_017005756.1 | XP_016861245.1 | 5 | 4449 | |||
XM_017005758.1 | XP_016861247.1 | 6 | 4567 | |||
GenBank mRNA | BC065764.1 | - | 5 | 357 | ||
BC094696.1 | - | 7 | 869 | 30529418 |