Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010863.2 | NP_001010863.1 | 4, 5 | 341 | ||
NM_001288740.1 | NP_001275669.1 | 5, 6 | 420 | |||
NM_001288743.1 | NP_001275672.1 | 5, 6 | 498 | |||
XM_011539378.2 | XP_011537680.1 | 5, 6 | 678 | |||
XM_011539379.1 | XP_011537681.1 | 5, 6 | 678 | |||
XM_011539380.1 | XP_011537682.1 | 5, 6 | 678 | |||
XM_011539382.1 | XP_011537684.1 | 4, 5 | 521 | |||
XM_011539383.2 | XP_011537685.1 | 4, 5 | 521 | |||
XM_011539384.1 | XP_011537686.1 | 4, 5 | 521 | |||
XM_017015792.1 | XP_016871281.1 | 5, 6 | 678 | |||
XM_017015793.1 | XP_016871282.1 | 5, 6 | 600 | |||
XM_017015794.1 | XP_016871283.1 | 4, 5 | 521 | |||
GenBank mRNA | AL524299.3 | - | 5, 6 | 395 | ||
BC031641.1 | - | 4, 5 | 872 | |||
BC037922.2 | - | 5, 6 | 489 | |||
BC042857.1 | - | 5, 6 | 489 | |||
BC047724.1 | - | 3, 4 | 1369 | |||
BC104438.1 | - | 4, 5 | 303 | |||
BC104439.1 | - | 4, 5 | 303 | |||
DQ884399.1 | - | 5, 6 | 434 |