Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133475.1 | NP_597732.1 | 8 | 779 | ||
XM_011527756.2 | XP_011526058.1 | 9 | 689 | |||
XM_011527757.2 | XP_011526059.1 | 8 | 937 | |||
XM_011527758.2 | XP_011526060.1 | 8 | 602 | |||
XM_011527761.1 | XP_011526063.1 | 10 | 759 | |||
XM_011527764.2 | XP_011526066.1 | 7 | 850 | |||
XM_011527765.2 | XP_011526067.1 | 7 | 503 | |||
XM_017026438.1 | XP_016881927.1 | 10 | 750 | |||
XM_017026439.1 | XP_016881928.1 | 9 | 672 | |||
XM_017026440.1 | XP_016881929.1 | 8 | 645 | |||
GenBank mRNA | AB075861.1 | - | 6 | 476 | ||
BC028236.1 | - | 6 | 421 | |||
BI914435.1 | - | 6 | 473 |