Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080421.2 | NP_001073890.2 | 10 | 1112 | ||
XM_011527810.2 | XP_011526112.1 | 9 | 1227 | |||
XM_011527811.2 | XP_011526113.1 | 10 | 1257 | |||
XM_017026502.1 | XP_016881991.1 | 10 | 1257 | |||
GenBank mRNA | AB028955.2 | - | 10 | 1110 |