Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015030.1 | NP_055845.1 | 4, 5 | 728 | ||
XM_005248090.4 | XP_005248147.1 | 4, 5 | 741 | |||
XM_011513680.2 | XP_011511982.1 | 1, 2 | 203 | |||
XM_011513681.2 | XP_011511983.1 | 1, 2 | 203 | |||
XM_011513682.2 | XP_011511984.1 | 1, 2 | 203 | |||
XM_011513683.2 | XP_011511985.1 | 3, 4 | 577 | |||
XM_011513685.2 | XP_011511987.1 | 1, 2 | 203 | |||
XM_017008046.1 | XP_016863535.1 | 5, 6 | 1053 | |||
GenBank mRNA | AK131250.1 | - | 2, 3 | 403 | ||
AL833444.1 | - | 4, 5 | 688 |