Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080424.1 | NP_001073893.1 | 8 | 1078 | ||
XM_005256549.3 | XP_005256606.1 | 9 | 1230 | |||
XM_005256550.4 | XP_005256607.1 | 8 | 1072 | |||
XM_005256551.3 | XP_005256608.1 | 8 | 1111 | |||
XM_005256552.4 | XP_005256609.1 | 8 | 2084 | |||
XM_006721483.3 | XP_006721546.1 | 9 | 1175 | |||
XM_011523750.2 | XP_011522052.1 | 9 | 2435 | |||
XM_011523752.2 | XP_011522054.1 | 8 | 1075 | |||
XM_017024381.1 | XP_016879870.1 | 8 | 2521 | |||
GenBank mRNA | AB002344.2 | - | 8 | 1092 |