Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015123.2 | NP_055938.1 | 12, 13 | 1272 | ||
XM_005264722.1 | XP_005264779.1 | 12, 13 | 949 | |||
XM_005264723.1 | XP_005264780.1 | 2, 3 | 431 | |||
XM_017005989.1 | XP_016861478.1 | 12, 13 | 1056 | |||
XM_017005990.1 | XP_016861479.1 | 12, 13 | 949 | |||
XM_017005991.1 | XP_016861480.1 | 13, 14 | 1109 | |||
XM_017005992.1 | XP_016861481.1 | 12, 13 | 889 | |||
XM_017005993.1 | XP_016861482.1 | 12, 13 | 880 | |||
XM_017005994.1 | XP_016861483.1 | 12, 13 | 2722 | |||
XM_017005995.1 | XP_016861484.1 | 6, 7 | 466 | |||
XM_017005996.1 | XP_016861485.1 | 2, 3 | 168 | |||
XM_017005997.1 | XP_016861486.1 | 2, 3 | 431 | |||
GenBank mRNA | AB023230.1 | - | 12, 13 | 1041 | ||
AK295045.1 | - | 7, 8 | 514 | |||
AL832231.1 | - | 2, 3 | 269 | |||
DC340455.1 | - | 7, 8 | 483 |